Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Ammonia 500 ppm | 7664-41-7 | MFCD00011418 | 500 mL
SDP

Catalog No. NC1107797
Encompass_Preferred

Ammonia 500 ppm | Mol Wt: 17.03 | 7664-41-7 | MFCD00011418 | 500 mL

Catalog No. NC1107797 Supplier Aqua Solutions Supplier No. 0370500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonia 500 ppm | Mol Wt: 17.03 | 7664-41-7 | MFCD00011418 | 500 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.