Learn More
Hach Company Ammonia TNTplus Vial Test, HR (2-47 mg/L NH₃-N), 25 Tests
Catalog No. 501998127 Shop All Hach Company Products
Encompass
Ammonia TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Ammonia Nitrogen by the Salicylate Method. EPA compliant.
Catalog No. 50-199-8127 Supplier Hach Company Supplier No. TNT832
Ammonia TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Ammonia Nitrogen by the Salicylate Method. EPA compliant.