Learn More
Hach Company Ammonia TNTplus Vial Test, UHR+ (100 - 1800 mg/L NH₃-N), 25 Tests
Catalog No. 501998129 Shop All Hach Company Products
Encompass
For samples with very high Ammonia concentrations - ideal for monitoring Ammonia concentrations during the Anaerobic Ammonium Oxidation (Anammox) process. Ammonia TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. For determination of very high Ammonia concentrations, e.g. in centrate, filtrate or press water by the Salicylate method. With Ammonia TNTplus reagents, simply add sample and add the Dosicap zip reagent, shake the vial and insert into your Hach spectrophotometer! The Direct Read Technology will provide your Ammonia results in mg/L. No more sample dilutions, cleaning glassware, dealing with large amounts of waste, and worrying about human errors.25 Ammonia vials per box.
Catalog No. 50-199-8129 Supplier Hach Company Supplier No. TNT834
For samples with very high Ammonia concentrations - ideal for monitoring Ammonia concentrations during the Anaerobic Ammonium Oxidation (Anammox) process. Ammonia TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. For determination of very high Ammonia concentrations, e.g. in centrate, filtrate or press water by the Salicylate method. With Ammonia TNTplus reagents, simply add sample and add the Dosicap zip reagent, shake the vial and insert into your Hach spectrophotometer! The Direct Read Technology will provide your Ammonia results in mg/L. No more sample dilutions, cleaning glassware, dealing with large amounts of waste, and worrying about human errors.25 Ammonia vials per box.