Learn More
Hach Company Ammonia TNTplus Vial Test, High Range 10 to 100 mg/L NH3-N, 25 tests, 0.1 mL sample, Salicylate method, Reaction pH 12.6, 15 min
Catalog No. NC1992813 Shop All Hach Company Products
Encompass
Ammonia TNTplus Vial Test is a colorimetric vial test for precise determination of high-range ammonia nitrogen in water and wastewater. Each kit contains 25 vials with pre-dosed, freeze-dried reagents in sealed caps and incorporates vial-level identification and lot-specific calibration to reduce handling, contamination, and analytical errors while improving repeatability and quality tracking.
  • Range 10 to 100 mg/L NH3-N for high-range ammonia monitoring.
  • 25 tests per kit.
  • Sample size 0.1 mL.
  • Integrated barcode on each vial.
  • Ten measurements around the vial to detect outliers and average results for improved accuracy.
  • 2D barcode and RFID tracking for batch, expiry and certificate of analysis information.
  • Reaction pH 12.6 and 15 minute reaction time to final absorbance.
  • Suitable for monitoring municipal and industrial wastewater inlets.
  • Catalog No. NC1992813 Supplier Hach Company Supplier No. TNT837
     Restricted Item
    Add to Cart
    Add to Cart
    Ammonia TNTplus Vial Test is a colorimetric vial test for precise determination of high-range ammonia nitrogen in water and wastewater. Each kit contains 25 vials with pre-dosed, freeze-dried reagents in sealed caps and incorporates vial-level identification and lot-specific calibration to reduce handling, contamination, and analytical errors while improving repeatability and quality tracking.
  • Range 10 to 100 mg/L NH3-N for high-range ammonia monitoring.
  • 25 tests per kit.
  • Sample size 0.1 mL.
  • Integrated barcode on each vial.
  • Ten measurements around the vial to detect outliers and average results for improved accuracy.
  • 2D barcode and RFID tracking for batch, expiry and certificate of analysis information.
  • Reaction pH 12.6 and 15 minute reaction time to final absorbance.
  • Suitable for monitoring municipal and industrial wastewater inlets.
  • Product Title