Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics Ammonia VACUettes Refill Kit (Range: 0 to 120 and 120 to 1200 ppm)
SDP

Catalog No. 13299522
Change view
Click to view available options
Range:
0 to 120 and 120 to 1200 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
13-299-522 0 to 120 and 120 to 1200 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains mercury. Dispose according to local, state or federal laws.
Catalog No. 13-299-522 Supplier CHEMetrics Supplier No. R1501B/DEL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Contains mercury. Dispose according to local, state or federal laws.

This product contains 30 self-filling VACUettes ampules to refill kit K-1510B for the analysis of ammonia. R-1501B is compatible with C-1501B and C-1510B comparators.

Specifications

Analytes Ammonia
Format Liquid
For Use With (Equipment) C-1501B and C-1510B comparator
Includes 30 self-filling VACUettes ampoules to refill kit K-1510B
Range 0 to 120 and 120 to 1200 ppm
Type Ammonia Refill
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Warning! Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.