Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics Ammonia VACUettes Refill Kit (Range: 0 to 60 and 60 to 600 ppm)
SDP

Catalog No. 13299508A
Change view
Click to view available options
Range:
0 to 60 and 60 to 600 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
13-299-508A 0 to 60 and 60 to 600 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains mercury. Dispose according to local, state or federal laws.
Catalog No. 13-299-508A Supplier CHEMetrics Supplier No. R1501A/DEL
Contains mercury. Dispose according to local, state or federal laws.

This product contains 30 self-filling VACUettes ampules to refill kit K-1510A for the analysis of ammonia. R-1501A is compatible with C-1501A and C-1510A comparators.

Specifications

Analytes Ammonia
Format Liquid
For Use With (Equipment) C-1501A and C-1510A comparators
Includes 30 self-filling VACUettes ampoules to refill kit K-1510A
Range 0 to 60 and 60 to 600 ppm
Type Ammonia Refill
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Warning! Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.