Learn More
Hach Company Ammonia Vial Test ULR
Catalog No. 5000207043 Shop All Hach Company Products
Encompass_Preferred
Ammonia Vial Test-ULR (0 005-0 050 mg/L NH -N)-20 TestsAmmonia HPT chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results everytime For determination of very low ammonia concentrations e g in wastewater effluents surface water drinking water samples or fish farming applications by the Salicylate Method Hach Method 10205 Convenient handling using 50 mm rectangular vials 20 ULR ammonia vials per box
Catalog No. 50-002-07043 Supplier Hach Company Supplier No. HPT383
Ammonia Vial Test-ULR (0 005-0 050 mg/L NH -N)-20 TestsAmmonia HPT chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results everytime For determination of very low ammonia concentrations e g in wastewater effluents surface water drinking water samples or fish farming applications by the Salicylate Method Hach Method 10205 Convenient handling using 50 mm rectangular vials 20 ULR ammonia vials per box