Learn More
Electron Microscopy Sciences Ammonia Water 0.3% 500 ML
SDP
Catalog No. 5031902
Encompass_Preferred
Used in Harris Hematoxylin (1900), Nuclei. References: Ehrlich, P., Biol. Centralbl., 6:214, 1886, Mallory, F.B., Pathological Technique, W.B. Saunders Co Philadelphia, p 73, 1938, Clark G., (ed.) Staining Procedures, 3rd edition, Williams & Wilkins, Baltimore, c. 1973, pp 33-34, and Luna, L.G., (Ed.), Manual of Histologic Staining Methods of the AFIP, 3rd ed, McGraw-Hill, NY, pp35 and 38, c 1968
Catalog No. 50-319-02 Supplier Electron Microscopy Sciences Supplier No. 267544A
Used in Harris Hematoxylin (1900), Nuclei. References: Ehrlich, P., Biol. Centralbl., 6:214, 1886, Mallory, F.B., Pathological Technique, W.B. Saunders Co Philadelphia, p 73, 1938, Clark G., (ed.) Staining Procedures, 3rd edition, Williams & Wilkins, Baltimore, c. 1973, pp 33-34, and Luna, L.G., (Ed.), Manual of Histologic Staining Methods of the AFIP, 3rd ed, McGraw-Hill, NY, pp35 and 38, c 1968