Learn More
Electron Microscopy Sciences Ammonia Water 0.3% 500 ML
SDP
Catalog No. 5031743
Encompass_Preferred
Used in McManus' Method (PAS) for Glycogen, McManus (1948). McManus, JFA: Stain Tech.; 23:99 (1948), AFIP Manual of Histologic Staining Techniques; L.G. Luna, 3rd Ed., New York: McGraw-Hill Publications, c. 1968, p. 160, or Mowry, RW: Annals of the New York Academy of Science, 106:402 (1963).
Catalog No. 50-317-43 Supplier Electron Microscopy Sciences Supplier No. 2631205
Used in McManus' Method (PAS) for Glycogen, McManus (1948). McManus, JFA: Stain Tech.; 23:99 (1948), AFIP Manual of Histologic Staining Techniques; L.G. Luna, 3rd Ed., New York: McGraw-Hill Publications, c. 1968, p. 160, or Mowry, RW: Annals of the New York Academy of Science, 106:402 (1963).