Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammoniu mLactate Solution

Catalog No. 377228100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-722-8100ML 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-722-8100ML Supplier Sigma Aldrich Supplier No. 377228100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammoniu mLactate is an ammonium salt of lactic acid. It is generally used as a feedstock for ethyl lactate and as a component of moisturizers.Ammoniu mLactate solution has been used in the ammoniu mLactate fermentation broth for the concentration of organic acid salts by conventional electrodialysis (EDC).

Specifications

CAS 7047-84-9
Molecular Formula C18H37AlO4
Refractive Index n20/D 1.363
Linear Formula CH3(CH2)16COOAl(OH)2
MDL Number MFCD00019932
Quantity 1 kg
Molecular Weight (g/mol) 344.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.