Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium 1-Pyrrolidinecarbodithioate, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002351 Shop All Avantor Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-002-351 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-002-351 Supplier Avantor Supplier No. B33702
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Use in Atomic Absorption Spectrophotometry

Chemical Identifiers

CAS 5108-96-3
Molecular Formula C5H8NS2-
Molecular Weight (g/mol) 146.246
InChI Key VSWDORGPIHIGNW-UHFFFAOYSA-M
PubChem CID 4311638
IUPAC Name pyrrolidine-1-carbodithioate
SMILES C1CCN(C1)C(=S)[S-]

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 10 g
Formula Weight 164.29
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Ammonium 1-Pyrrolidinecarbodithioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.