Learn More
Waters Corp Waters Corporation Mobile Phase, Ammonium Acetate 1M pH 5, 2 units, 10 mL
Catalog No. 502037726 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product, Ammonium Acetate 1M pH 5, is an HPLC standard and mobile phase designed for pesticide screening applications. It is supplied in two 10 mL ampules and is suitable for use in experiment setups that require a reverse-phase mobile phase.
  • Designed for pesticide screening applications.
  • Two 10 mL ampules of Ammonium Acetate, 1M pH 5.0.
  • Functions as an HPLC standard.
  • Utilizes reversed-phase separation mode.
  • Compatible with MS detection methods.
  • Liquid formulation.
  • Stores at room temperature.
  • Has a shelf life of 12 months.
  • Ships under ambient conditions.
Catalog No. 50-203-7726 Supplier Waters Corp Supplier No. 186006693
This product, Ammonium Acetate 1M pH 5, is an HPLC standard and mobile phase designed for pesticide screening applications. It is supplied in two 10 mL ampules and is suitable for use in experiment setups that require a reverse-phase mobile phase.
  • Designed for pesticide screening applications.
  • Two 10 mL ampules of Ammonium Acetate, 1M pH 5.0.
  • Functions as an HPLC standard.
  • Utilizes reversed-phase separation mode.
  • Compatible with MS detection methods.
  • Liquid formulation.
  • Stores at room temperature.
  • Has a shelf life of 12 months.
  • Ships under ambient conditions.