Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium acetate, ≥99.0% (calc. on dry substance, T), MilliporeSigma™ Supelco™

Catalog No. 111018829 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8829 25 g
11-101-8073 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8829 Supplier MilliporeSigma Supelco Supplier No. 7359425GF
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium acetate exists as a white crystal. It is prepared from acetic acid and ammonia by neutralization to pH 7.0. It is widely used in analytical chemistry, drugs and in textile dyeing.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00013066

Specifications

pH 6.2 to 7.5 (25°C, 1 M aq. soln.)
Melting Point 110°C to 112°C
CAS 631-61-8
Quantity 25 g
Physical Form Solid
Linear Formula CH3 CO2 NH4
Solubility Information Water: 1 M (at 20°C, clear, colorless)
Formula Weight 77.08
Percent Purity ≥99.0% (calc. on dry substance, T)
Grade Mass Spec
RTECSNumber AF3675000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.