Learn More
Strem, An Ascensus Company CAS# 631-61-8. 500g. Ammonium acetate, anhydrous, 97+% (ACS). MFCD00013066
SDP
Catalog No. 5090113560
Encompass
CAS# 631-61-8. 500g. Ammonium acetate, anhydrous, 97+% (ACS). MFCD00013066. Molecular Weight: 77.08. Molecular Formula: NH4OOCCH3. Color/form: white xtl. Strem# 93-0201. http://www.strem.com/catalog/v/93-0201/
Catalog No. 50-901-13560 Supplier Strem, An Ascensus Company Supplier No. 930201500G
CAS# 631-61-8. 500g. Ammonium acetate, anhydrous, 97+% (ACS). MFCD00013066. Molecular Weight: 77.08. Molecular Formula: NH4OOCCH3. Color/form: white xtl. Strem# 93-0201. http://www.strem.com/catalog/v/93-0201/