Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Acetate Buffer pH 4.2 - 4.6 APHA For Iron Analysis (1L)
SDP

Catalog No. NC9284076
Encompass_Preferred

Acetate Buffer pH 4.2 - 4.6 APHA For Iron Analysis (1L)

Catalog No. NC9284076 Supplier Aqua Solutions Supplier No. 04201L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetate Buffer pH 4.2 - 4.6 APHA For Iron Analysis (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.