Learn More
Description
- Suitable for EPA 1633, EPA 533, EPA 537.1
- HPLC, LC/MS, ion chromatography suitable
Specifications
Specifications
| CAS | 631-61-8 |
| Quantity | 50 g |
| Molecular Weight (g/mol) | 77.08 |
| Percent Purity | ≥98.0% (Acidimetric) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.