Learn More
Crescent Chemical Co Inc AMMONIUM ACETATE MOL BIO GRADE
SDP
Catalog No. 50248428
Encompass_Preferred
Ammonium acetate molecular biology grade Ammonium acetate can be used instead of sodium acetate for precipitation of nucleic acids. In general, for precipitation of DNA a 7.5 M stock solution and for RNA a 3 M stock solution are used. DNase/RNase not detected. /
Catalog No. 50-248-428 Supplier Crescent Chemical Co Inc Supplier No. SE39750.01
Ammonium acetate molecular biology grade Ammonium acetate can be used instead of sodium acetate for precipitation of nucleic acids. In general, for precipitation of DNA a 7.5 M stock solution and for RNA a 3 M stock solution are used. DNase/RNase not detected. /