Learn More
Crescent Chemical Co Inc AMMONIUM ACETATE SOLUTION 7.5M
SDP
Catalog No. 50248430
Encompass_Preferred
Ammonium acetate solution, 7.5 M molecular biology grade 7.5 M stock solution (ammonium acetate, cat. no. 39750: 578.1 g/l) for precipitation of DNA. DNase/RNase not detected.Short oligonucleotides and free dNTPs do not coprecipitate with DNA when precipitated with NH4OAc. Two co
Catalog No. 50-248-430 Supplier Crescent Chemical Co Inc Supplier No. SE39751.02
Ammonium acetate solution, 7.5 M molecular biology grade 7.5 M stock solution (ammonium acetate, cat. no. 39750: 578.1 g/l) for precipitation of DNA. DNase/RNase not detected.Short oligonucleotides and free dNTPs do not coprecipitate with DNA when precipitated with NH4OAc. Two co