Learn More
Crescent Chemical Co Inc Ammonium Atate Solution, 631-61-8, MFCD00013066, 250mL
SDP
Catalog No. 50248429
Encompass_Preferred
Molecular Formula C2H4O2.H3N, Molecular Weight 77.082, Molecular Biology Grade, 7.5 M stock solution (ammonium acetate, cat. no. 39750: 578.1 g/l) for precipitation of DNA. DNase/RNase not detected.Short oligonucleotides and free dNTPs do not coprecipitate with DNA when precipitated with NH4OAc. Two consecutive precipitation steps are enough to remove 99 % of free dNTPs.
Catalog No. 50-248-429 Supplier Crescent Chemical Co Inc Supplier No. SE39751.01
Molecular Formula C2H4O2.H3N, Molecular Weight 77.082, Molecular Biology Grade, 7.5 M stock solution (ammonium acetate, cat. no. 39750: 578.1 g/l) for precipitation of DNA. DNase/RNase not detected.Short oligonucleotides and free dNTPs do not coprecipitate with DNA when precipitated with NH4OAc. Two consecutive precipitation steps are enough to remove 99 % of free dNTPs.