Learn More
Medchemexpress LLC Ammonium aluminum sulfate dodecahydrate | 7784-26-1 | 25 G
SDP
Catalog No. 5000405627
Encompass_Preferred
Ammonium aluminum sulfate dodecahydrate is a biomolecule precipitation agent that can change the solubility of proteins by interacting with water molecules, thereby effectively purifying proteins. It is for research use only.
  • Biomolecule precipitation agent
  • Changes solubility of proteins
  • Effectively purifies proteins
  • For research use only
Catalog No. 50-004-05627 Supplier Medchemexpress LLC Supplier No. 25GHYB0560A
Add to Cart
Add to Cart
Ammonium aluminum sulfate dodecahydrate is a biomolecule precipitation agent that can change the solubility of proteins by interacting with water molecules, thereby effectively purifying proteins. It is for research use only.
  • Biomolecule precipitation agent
  • Changes solubility of proteins
  • Effectively purifies proteins
  • For research use only