Learn More
Medchemexpress LLC Ammonium aluminum sulfate dodecahydrate | 7784-26-1 | 250 G
SDP
Catalog No. 5000407439
Encompass_Preferred
Ammonium aluminum sulfate dodecahydrate is a biomolecule precipitation agent. It changes the solubility of proteins by interacting with water molecules, thereby effectively purifying proteins. For research use only.
  • Biomolecule precipitation agent
  • Changes solubility of proteins
  • Interacts with water molecules
  • Effectively purifies proteins
  • For research use only
Catalog No. 50-004-07439 Supplier Medchemexpress LLC Supplier No. 250GHYB0560A
Ammonium aluminum sulfate dodecahydrate is a biomolecule precipitation agent. It changes the solubility of proteins by interacting with water molecules, thereby effectively purifying proteins. For research use only.
  • Biomolecule precipitation agent
  • Changes solubility of proteins
  • Interacts with water molecules
  • Effectively purifies proteins
  • For research use only