Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium aluminum sulfate500G
Catalog No. 502897428
Encompass
Ammonium aluminum sulfate dodecahydrate commonly called as ammonium alum is widely used in the water purification cosmetic formulation fertilizers and food preservation. Its thermal decomposition has been studied. It has high latent heat of 251kJkg-1 with a potential use in the heating system. Its utility as a flame-retardant for cotton fabric has been studied.
Catalog No. 50-289-7428 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A2140500G
Ammonium aluminum sulfate dodecahydrate commonly called as ammonium alum is widely used in the water purification cosmetic formulation fertilizers and food preservation. Its thermal decomposition has been studied. It has high latent heat of 251kJkg-1 with a potential use in the heating system. Its utility as a flame-retardant for cotton fabric has been studied.