Learn More
Ethos Biosciences Ammonium Hydroxide, ACS, 1gal, Ethos
Catalog No. 5000574171
Encompass_Preferred
Ammonium Hydroxide is a clear, aqueous ammonia solution suitable for biological, general laboratory, and analytical applications. Commonly used for pH adjustment, reagent preparation, and buffer formulation, it supports a variety of routine laboratory procedures. It is also used in staining protocols, sample preparation, and other biological workflows requiring an alkaline solution. The solution provides consistent concentration for reliable and reproducible results. Supplied in chemically compatible containers, Ammonium Hydroxide is appropriate for clinical, research, pharmaceutical, and educational laboratory environments.
Catalog No. 50-005-74171 Supplier Ethos Biosciences Supplier No. 3535G
Ammonium Hydroxide is a clear, aqueous ammonia solution suitable for biological, general laboratory, and analytical applications. Commonly used for pH adjustment, reagent preparation, and buffer formulation, it supports a variety of routine laboratory procedures. It is also used in staining protocols, sample preparation, and other biological workflows requiring an alkaline solution. The solution provides consistent concentration for reliable and reproducible results. Supplied in chemically compatible containers, Ammonium Hydroxide is appropriate for clinical, research, pharmaceutical, and educational laboratory environments.