Learn More
Medchemexpress LLC Ammonium benzoate | 1863-63-4 | 100g
SDP
Catalog No. 5000300672
Encompass_Preferred
Ammonium benzoate is the ammonium salt of benzoic acid which is used as a preservative in some adhesives and rubber emulsions and is also a commonly used chemical synthesis intermediate
Catalog No. 50-003-00672 Supplier Medchemexpress LLC Supplier No. HYW015761100G
Ammonium benzoate is the ammonium salt of benzoic acid which is used as a preservative in some adhesives and rubber emulsions and is also a commonly used chemical synthesis intermediate