Learn More
Medchemexpress LLC Ammonium benzoate | 1863-63-4 | MFCD00013069 | 98.0% | 139.15 g/mol | C7H9NO2 | 500 G
SDP
Catalog No. 5000300673
Encompass_Preferred
Ammonium benzoate is the ammonium salt of benzoic acid commonly used as a preservative in some adhesives and rubber emulsions and as an intermediate in chemical synthesis. It appears as a white crystalline solid with formula C7H9NO2 and molecular weight 139.15 g/mol, and is typically supplied in laboratory reagent pack sizes for research and industrial use.
  • Ammonium salt of benzoic acid with CAS 1863-63-4.
  • Used as a preservative in adhesives and rubber emulsions.
  • Serves as a chemical synthesis intermediate.
  • White crystalline solid, soluble in water.
  • Supplied in laboratory-scale packaging suitable for research.
  • Molecular formula C7H9NO2; molecular weight 139.15 g/mol.
Catalog No. 50-003-00673 Supplier Medchemexpress LLC Supplier No. HYW015761500G
Ammonium benzoate is the ammonium salt of benzoic acid commonly used as a preservative in some adhesives and rubber emulsions and as an intermediate in chemical synthesis. It appears as a white crystalline solid with formula C7H9NO2 and molecular weight 139.15 g/mol, and is typically supplied in laboratory reagent pack sizes for research and industrial use.
  • Ammonium salt of benzoic acid with CAS 1863-63-4.
  • Used as a preservative in adhesives and rubber emulsions.
  • Serves as a chemical synthesis intermediate.
  • White crystalline solid, soluble in water.
  • Supplied in laboratory-scale packaging suitable for research.
  • Molecular formula C7H9NO2; molecular weight 139.15 g/mol.