Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium biborate tetrahydrate

Catalog No. A4380500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4380500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A4380500G Supplier Sigma Aldrich Supplier No. A4380500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium biborate tetrahydrate

Specifications

CAS 12228-87-4
Quantity 500 g
Molecular Formula H8B4N2O7 -+ 4H2O
Linear Formula (NH4)2B4O7 -+ 4H2O
MDL Number MFCD00149601
Synonym Ammonium tetraborate tetrahydrate
Molecular Weight (g/mol) 263.38
Percent Purity 98.9-101.2%B2O3 basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.