Learn More
ALADDIN SCIENTIFIC CORPORATION Ammonium bicarbonate, 100g
Catalog No. 5000082528
Encompass_Preferred
Ammonium bicarbonate is a commonly used reagent for industrial and research procedures Ammonium bicarbonate is volatile in solution and releases ammonia and CO2 This property makes ammonium bicarbonate a good buffer for such applications as lyophilization and matrix assisted laser desorption Ammonium bicarbonate is also utilized for the in-gel digestion of proteins by trypsin and in the MALDI mass spectrometric analysis of proteins A buffer applications such as lyophilization and matrix assisted laser desorption
Catalog No. 50-000-82528 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. A110537100G
Ammonium bicarbonate is a commonly used reagent for industrial and research procedures Ammonium bicarbonate is volatile in solution and releases ammonia and CO2 This property makes ammonium bicarbonate a good buffer for such applications as lyophilization and matrix assisted laser desorption Ammonium bicarbonate is also utilized for the in-gel digestion of proteins by trypsin and in the MALDI mass spectrometric analysis of proteins A buffer applications such as lyophilization and matrix assisted laser desorption