Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium bicarbonate | 1066-33-7 | MFCD00012138 | 500 g
Catalog No. NC1471744
Encompass
Ammonium bicarbonate | Purity: 99% | Mol Wt: 79.06 | 1066-33-7 | MFCD00012138 | 500 g
Catalog No. NC1471744 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A6141500G
Ammonium bicarbonate | Purity: 99% | Mol Wt: 79.06 | 1066-33-7 | MFCD00012138 | 500 g