Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bicarbonate, Fisher BioReagents

Catalog No. BP2413500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2413500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP2413500

missing translation for 'validMainframeMsg'

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol

Poly Bottle; 500g

Ammonium Bicarbonate is used for many biochemical applications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Bicarbonate
Melting Point 105°C
Color White
CAS 1066-33-7
pH 8
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 21.30 to 21.73 %
Molecular Formula CH5NO3
Linear Formula NH4HCO3
MDL Number MFCD00012138
Merck Index 15, 494
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
SMILES N.OC(O)=O
IUPAC Name carbonic acid amine
Molecular Weight (g/mol) 79.06
PubChem CID 14013
Formula Weight 79.06
Percent Purity 21.30 to 21.73%
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 67 hPa at 20°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.