Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bifluoride, Technical, Ricca Chemical
SDP

Catalog No. RDCA0420500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCA0420500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCA0420500 Supplier Ricca Chemical Company Supplier No. RDCA0420500B1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium bifluoride in a technical grade, supplied as a solid.

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Physical Form Fine Crystals or Crystalline Powder
CAS Max % 100
Quantity 500 g
Identification Pass
Packaging White Poly Wide Mouth Bottle
Grade Laboratory
Chemical Name or Material Ammonium Bifluoride
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.