Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bisulfate, Purum p.a., ≥98.0% (T), Solstice

Catalog No. 60044620 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-620 1 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-620 Supplier Solstice Supplier No. 098481KG

missing translation for 'validMainframeMsg'

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-63-6
Molecular Formula H5NO4S
Molecular Weight (g/mol) 115.10
MDL Number MFCD00051073
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
Synonym ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4
PubChem CID 24655
IUPAC Name ammonium hydrogen sulfate
SMILES [NH4+].OS([O-])(=O)=O

Specifications

Boiling Point 350°C
Melting Point 121°C to 145°C
Color Colorless
pH 1 (100g/L at 20°C)
Physical Form Crystalline
Quantity 1 kg
Linear Formula (NH4)HSO4
UN Number UN2506
Solubility Information Soluble in water, 0.5g in 10mL
Formula Weight 115.11g/mol
Percent Purity ≥98.0% (T)
Grade purum p.a.
Packaging Poly bottle
Chemical Name or Material Ammonium Bisulfate
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H314
DOTInformation 8/PGII
EINECSNumber 232-265-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.