Learn More
Ammonium bromide, ACS, 99.0% min
Description
Corrosion inhibitors, manufacturing of photographic films, plates, paper, lithography, fireproofing of woodsAmmonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 452° (sublimation) |
| Color | Colorless |
| Physical Form | Solid |
| Quantity | 500 g |
| Assay Percent Range | 99% min. |
| Solubility Information | Freely soluble in water,methanol,ethanol,and acetone. Slightly soluble in ether. Practically insoluble in ethyl acetateSoluble in water,methanol,ethanol and acetone. Slightly soluble in ether. |
| Formula Weight | 97.97 |
| Grade | ACS |
| Sensitivity | Hygroscopic |
| Density | 2.429 |
| Show More |
Safety and Handling
| Hazard Category | H319 |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 235-183-8 |
| RTECSNumber | BO9155000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.