Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium bromide ACS reage500G
Catalog No. 502898864
Encompass
Laser excited Raman spectral study reveals that 1?0 torsional transitions of the NH4 ion in phase III of NH4Br were observed at 331cm-1. Neutron diffraction studies have been conducted to evaluate the P-T phase diagram of NH4Br and its deuterated form (ND3Br).
Catalog No. 50-289-8864 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 213349500G
Add to Cart
Add to Cart
Laser excited Raman spectral study reveals that 1?0 torsional transitions of the NH4 ion in phase III of NH4Br were observed at 331cm-1. Neutron diffraction studies have been conducted to evaluate the P-T phase diagram of NH4Br and its deuterated form (ND3Br).