Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bromide, ACS Reagent, ≥99.0%, Solstice

Catalog No. 60045224 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-224 500 g Poly Bottle
60-045-223 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-224 Supplier Solstice Supplier No. 213349500G

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Melting Point 452°C
Color Colorless
pH 5 to 6.5 (at 50g/L 25°C)
Quantity 500 g
Linear Formula NH4Br
UN Number NONH for all modes of transport
Formula Weight 97.94g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Residue after Ignition ≤0.01%
Appearance Crystals
Vapor Pressure 1mmHg (198.3°C)
Density 2.43g/mL at 25°C
Chemical Name or Material Ammonium Bromide
Show More Show Less
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 235-183-8
RTECSNumber BO9155000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.