Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium bromide, Puratronic™, 99.999% (metals basis)

Catalog No. AA1298114
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

Ammonium bromide is used in pharmaceuticals for its bromide ion. It is used for a variety of analytical chemistry applications. It is also used for photography in films, plates and papers; in fireproofing of wood; in lithography and process engraving and as corrosion inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Melting Point 452°C (sublimation)
Color White
Physical Form Crystalline
Quantity 25 g
Merck Index 14,505
Solubility Information Soluble in water (970mg/ml at 25°C),methanol,ethanol,alcohol (freely),and acetone (freely).
Formula Weight 97.97
Percent Purity 99.999%
Grade Puratronic™
Sensitivity Hygroscopic
Density 2.429 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium bromide
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 235-183-8
RTECSNumber BO9155000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.