Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Carbonate TS, Ricca Chemical
SDP

$168.70 - $306.00

Specifications

CAS 506-87-6 , 7732-18-5 , 1336-21-6
Grade Laboratory
Packaging Natural Poly Bottle
pH Alkaline
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R0622500120
SDP
View Documents
Ricca Chemical Company
R0622500120A
120 mL
Each for $168.70
Add to Cart
 
6225-16
SDP
View Documents
Ricca Chemical Company
622.516
500 mL
Each for $306.00
Add to Cart
 
Description

Description

Specifications

Specifications

506-87-6, 7732-18-5, 1336-21-6
Laboratory
Natural Poly Bottle
Alkaline
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.