Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ammonium cerium(IV) sulfate dihydrate

Catalog No. 221759100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-175-9100G 100 g
22-175-9500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-175-9100G Supplier Sigma Aldrich Supplier No. 221759100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ammonium cerium(IV) sulfate dihydrate

Specifications

CAS 10378-47-9
Melting Point 130°C
Molecular Formula H16CeN4O16S4 -+ 2H2O
Linear Formula Ce(NH4)4(SO4)4 -+ 2H2O
MDL Number MFCD00149417
Quantity 100 g
Synonym Ceric ammonium sulfate; Cerium(IV) ammonium sulfate
Molecular Weight (g/mol) 632.55
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.