Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium cerium(IV) sulfate solution, Solstice

Catalog No. 6000328 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-003-28 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-003-28 Supplier Solstice Supplier No. 379411L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, Reag. Ph. Eur., 0.1 M Ce(NH4)4(SO4)4

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7642-09-3
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00148851
InChI Key ZQDPJFUHLCOCRG-WAYWQWQTSA-N
Synonym cis-3-hexene, z-3-hexene, 3-hexene, z, z-hex-3-ene, 3z-hex-3-ene, 3-hexene, 3z, unii-50181bz6yt, 3z-3-heptene, 3z-3-hexene #, 3-hexene cis
PubChem CID 643783
IUPAC Name (Z)-hex-3-ene
SMILES CCC=CCC

Specifications

Physical Form Liquid
Quantity 1 L
UN Number UN3264
Formula Weight 596.52g/mol
Grade Volumetric
Packaging Poly Bottle
Chemical Name or Material Ammonium cerium(IV) sulfate solution
Hazard Statement P305 + P351 + P338
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.