Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium ceriumIV nitrate500G
Catalog No. 502897519
Encompass
Ammonium cerium(IV) nitrate (Cerium(IV) ammonium nitrate CAN) is a versatile reagent for oxidative electron transfer reactions. It participates in various novel carbon-carbon bond-forming reactions involved in one-pot synthesis of dihydrofurans tetrahydrofurans and aminotetralins. Iodine/ammonium cerium(IV) nitrate has been employed in direct iodination of various ketones to afford the corresponding iodo ketones. CAN has been reported as an powerful one-electron oxidant. It also participates in various carbon-heteroatom bond-forming reactions.
Catalog No. 50-289-7519 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 22249500G
Add to Cart
Add to Cart
Ammonium cerium(IV) nitrate (Cerium(IV) ammonium nitrate CAN) is a versatile reagent for oxidative electron transfer reactions. It participates in various novel carbon-carbon bond-forming reactions involved in one-pot synthesis of dihydrofurans tetrahydrofurans and aminotetralins. Iodine/ammonium cerium(IV) nitrate has been employed in direct iodination of various ketones to afford the corresponding iodo ketones. CAN has been reported as an powerful one-electron oxidant. It also participates in various carbon-heteroatom bond-forming reactions.