Learn More
Medchemexpress LLC Ammonium chloride | 12125-02-9 | MFCD00143604 | 99.7% | 53.49 | NH4Cl | 1g
SDP
Catalog No. 5000267049
Encompass_Preferred
Ammonium chloride as a heteropolar compound with pH value regulation can cause intracellular alkalization and metabolic acidosis thus effecting enzymatic activity and influencing the process of biological system Ammonium chloride is an autophagy inhibitor Ammonium chloride is also a lysosome inhibitor[1][2][3]
Catalog No. 50-002-67049 Supplier Medchemexpress LLC Supplier No. HYY12691G
Ammonium chloride as a heteropolar compound with pH value regulation can cause intracellular alkalization and metabolic acidosis thus effecting enzymatic activity and influencing the process of biological system Ammonium chloride is an autophagy inhibitor Ammonium chloride is also a lysosome inhibitor[1][2][3]