Learn More
Medchemexpress LLC Ammonium chloride-15N | 39466-62-1 | MFCD00075417 | 98.0% | 54.48 | 15NH4Cl | 100mg
SDP
Catalog No. 5000267388
Encompass_Preferred
Ammonium chloride-15N is the 15N labeled Ammonium chloride (HY-Y1269) Ammonium chloride as a heteropolar compound with pH value regulation can cause intracellular alkalization and metabolic acidosis thus effecting enzymatic activity and influencing the process of biological system Ammonium chloride is an autophagy inhibitor Ammonium chloride is also a lysosome inhibitor[1][2][3]
Catalog No. 50-002-67388 Supplier Medchemexpress LLC Supplier No. HYY1269S100MG
Ammonium chloride-15N is the 15N labeled Ammonium chloride (HY-Y1269) Ammonium chloride as a heteropolar compound with pH value regulation can cause intracellular alkalization and metabolic acidosis thus effecting enzymatic activity and influencing the process of biological system Ammonium chloride is an autophagy inhibitor Ammonium chloride is also a lysosome inhibitor[1][2][3]