Learn More
Cambridge Isotope Laboratories Ammonium Chloride (15N, 99%), 39466-62-1, MFCD00075417, 10G
Catalog No. NC1313549
Encompass
Chemical Formula: NH4Cl, Molecular Weight: 54.48, Synonym: 15N Labeled ammonium chloride. Ammonium-15N chloride has been used: as a medium supplement for the incorporation of 15N in sporulation protein, CsfB, for NMR spectroscopy, in assaying nitrogen loss in marine sediment, and for the incorporation of 15N in apolipoprotein A-I for mass spectrum analysis.
Catalog No. NC1313549 Supplier Cambridge Isotope Laboratories Supplier No. NLM46710
Chemical Formula: NH4Cl, Molecular Weight: 54.48, Synonym: 15N Labeled ammonium chloride. Ammonium-15N chloride has been used: as a medium supplement for the incorporation of 15N in sporulation protein, CsfB, for NMR spectroscopy, in assaying nitrogen loss in marine sediment, and for the incorporation of 15N in apolipoprotein A-I for mass spectrum analysis.