Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium chloride, 12125-02-9, MFCD00011420, 500 G
Catalog No. NC0559163
Encompass
Linear Formula: NH4Cl, Molecular Weight: 53.49, ACS reagent, ≥99.5%, Synonym: Salmiac. Ammonium chloride (NH4Cl) has been used as one of the components of red blood cell (RBC) lysis buffer for use in various studies.
Catalog No. NC0559163 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 213330500G
Linear Formula: NH4Cl, Molecular Weight: 53.49, ACS reagent, ≥99.5%, Synonym: Salmiac. Ammonium chloride (NH4Cl) has been used as one of the components of red blood cell (RBC) lysis buffer for use in various studies.