Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium chloride, 12125-02-9, MFCD00011420, 500 G
Catalog No. NC0684400
Encompass
Linear Formula: NH4Cl, Molecular Weight: 53.49, for molecular biology, suitable for cell culture, ≥99.5%, Synonym: Salmiac. Ammonium chloride has been used: as an inhibitor of endosome acidification in vero cells, as an autophagy inhibitor in vascular smooth muscle cell (VSMC), and in the depletion and lysis of erythrocytes in testis parenchyma homogenate.
Catalog No. NC0684400 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A9434500G
Linear Formula: NH4Cl, Molecular Weight: 53.49, for molecular biology, suitable for cell culture, ≥99.5%, Synonym: Salmiac. Ammonium chloride has been used: as an inhibitor of endosome acidification in vero cells, as an autophagy inhibitor in vascular smooth muscle cell (VSMC), and in the depletion and lysis of erythrocytes in testis parenchyma homogenate.