Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride
SCI_ED

Catalog No. S70609 Shop All Fisherbrand Products
Change view
Click to view available options
Lab Grade - Granular; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S70609 Lab Grade - Granular; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S70609

missing translation for 'validMainframeMsg'

Freezing point depression

Specifications

Color White
CAS 12125-02-9
Quantity 500 g
Molecular Formula ClH4N
Linear Formula NH4Cl
MDL Number MFCD00011420
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC Name amine hydrochloride
Molecular Weight (g/mol) 53.49
PubChem CID 25517
ChEBI CHEBI:31206
Formula Weight 53.49
Packaging Lab Grade - Granular
Chemical Name or Material Ammonium Chloride
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife Indefinite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.