Learn More
Strem, An Ascensus Company CAS# 12125-02-9. 500g. Ammonium chloride, 99.5+% (ACS). MFCD00011420
SDP
Catalog No. 5090113565
Encompass
CAS# 12125-02-9. 500g. Ammonium chloride, 99.5+% (ACS). MFCD00011420. Molecular Weight: 53.49. Molecular Formula: NH4Cl. Color/form: white gran. Strem# 93-0208. http://www.strem.com/catalog/v/93-0208/
Catalog No. 50-901-13565 Supplier Strem, An Ascensus Company Supplier No. 930208500G
CAS# 12125-02-9. 500g. Ammonium chloride, 99.5+% (ACS). MFCD00011420. Molecular Weight: 53.49. Molecular Formula: NH4Cl. Color/form: white gran. Strem# 93-0208. http://www.strem.com/catalog/v/93-0208/