Learn More
Strem, An Ascensus Company CAS# 12125-02-9. 100g. Ammonium chloride (99.999%) PURATREM. MFCD00011420
SDP
Catalog No. 5090113580
Encompass
CAS# 12125-02-9. 100g. Ammonium chloride (99.999%) PURATREM. MFCD00011420. Molecular Weight: 53.49. Molecular Formula: NH4Cl. Color/form: white xtl. Strem# 93-0242. http://www.strem.com/catalog/v/93-0242/
Catalog No. 50-901-13580 Supplier Strem, An Ascensus Company Supplier No. 930242100G
CAS# 12125-02-9. 100g. Ammonium chloride (99.999%) PURATREM. MFCD00011420. Molecular Weight: 53.49. Molecular Formula: NH4Cl. Color/form: white xtl. Strem# 93-0242. http://www.strem.com/catalog/v/93-0242/