Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium chloride ACS reag
Catalog No. 502898033
Encompass
Crystals of ammonium chloride (NH4Cl) belong to the cubic crystal system. It can be synthesized by reacting HCl with aqueous ammonia SOLUTIONons. NH4Cl is formed as a by-product during soda manufactured by Solvay process.
Catalog No. 50-289-8033 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 21333012KG
Crystals of ammonium chloride (NH4Cl) belong to the cubic crystal system. It can be synthesized by reacting HCl with aqueous ammonia SOLUTIONons. NH4Cl is formed as a by-product during soda manufactured by Solvay process.