Learn More
Sigma Aldrich Fine Chemicals Biosciences Ammonium chloride ACS reag50KG
Catalog No. 502903917
Encompass
Crystals of ammonium chloride (NH4Cl) belong to the cubic crystal system. It can be synthesized by reacting HCl with aqueous ammonia SOLUTIONons. NH4Cl is formed as a by-product during soda manufactured by Solvay process.
Catalog No. 50-290-3917 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 21333050KG
Add to Cart
Add to Cart
Crystals of ammonium chloride (NH4Cl) belong to the cubic crystal system. It can be synthesized by reacting HCl with aqueous ammonia SOLUTIONons. NH4Cl is formed as a by-product during soda manufactured by Solvay process.