Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride/Ammonium Hydroxide Buffer (Copper Buffer), Reagents

Catalog No. 01335961
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-961 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-961 Supplier Reagents Holdings Llc Supplier No. CA1009001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Chloride/Ammonium Hydroxide Buffer (Copper Buffer), Reagent, use for EDTA titrations. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonium Chloride/Ammonium Hydroxide Buffer (Copper Buffer)
CAS 7732-18-5, 1336-21-6, 1336-21-6
Color Colorless
Concentration 1 M
For Use With (Application) Titration
pH 11.5 to 12.5
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.0185
Sterility Non-sterile
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.